Learn More
Cytiva Benchkote sheet for ÄKTA systems
Benchkote sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
20.15 € - 44.60 €
Specifications
Material Absorbent Paper, Laminated Polyethylene
Description For use with ÄKTA go
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16497645
View Documents
Cytiva
2300-10094
25/Pk.
44.60 €
Pack of 50
In Stock
to see stock.
Add to basket
16477645
View Documents
Cytiva
2300-10092
10/Pk.
20.15 €
Pack of 10
Estimated Shipment: 22-09-2025
to see stock.
Add to basket
16487645
View Documents
Cytiva
2300-10093
50/Pk.
28.75 €
Pack of 25
Estimated Shipment: 22-09-2025
to see stock.
Add to basket
Description
Description
Why Benchkote sheet for lab ÄKTA systems?
  • Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
  • Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
  • Smooth, absorbent Whatman™ paper quickly absorbs spills.
  • Laminated polyethylene layer prevents flow-through to the working surface.
  • Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
  • Convenient and easy to use.
  • Benchkote sheets are manufactured of absorbent, impermeable material designed to protect lab surfaces against hazardous spills.
  • The high-quality, smooth, and absorbent Whatman™ paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
Specifications
Specifications
Absorbent Paper, Laminated Polyethylene
For use with ÄKTA go
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cytiva Benchkote sheet for ÄKTA systems