Learn More
Gibco™ BenchStable™ MEM
Gibco Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media.
26.07 € - 246.00 €
Specifications
Content And Storage | Store at room temperature. Protect from light. |
---|---|
Cell Line | HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts |
Classification | Animal Origin-free |
Concentration | 1 X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16477060
|
Gibco™
A4192201 |
500 mL |
26.07 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16487060
|
Gibco™
A4192202 |
10 x 500 mL |
246.00 €
5000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
BenchStable MEM has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional MEM when supplemented with ≥10% fetal bovine serum (FBS).
This MEM is modified as follows:
• Optimized for room temperature stability
• With GlutaMAX, Phenol Red
• Without HEPES
Using BenchStable MEM
MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This MEM includes Earle's salts. MEM with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable MEM requires supplementation with ≥10% fetal bovine serum (FBS).
Specifications
Store at room temperature. Protect from light. | |
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
Low Glucose, GlutaMAX, Phenol Red | |
Energy efficient, Fewer resources used, Sustainable packaging | |
BenchStable™ |
HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts | |
1 X | |
MEM (Minimum Essential Medium) | |
Sterile-filtered | |
No HEPES, No Sodium Pyruvate | |
cGMP-compliant under the ISO 9001: 2015 standard | |
Room Temperature |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.