Learn More
Sartorius Benchtop Arium pro UF Ultrapure Water System
BIOPHARMAGREENER_CHOICE
Provides widest selection of features and options for customised configuration.
Brand:  Sartorius H2OPRO-UF-T
8750.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11785006
  • 8970.00 € / Each
Explore more special offers
Add to basket
Description
Description
  • Modular units pre-assembled according to specific applications
  • Lowest TOC level, along with real-time monitoring function
  • Efficient removal of endotoxins, DNase and RNase
  • High Flexibility
  • Total ease of operation
  • Meets and exceeds the ASTM Type 1 standard, ensuring reproducibility of results
  • Applications: HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC analysis, PCR, Electrophoresis, Endotoxin analysis, Immunocytochemistry, Pure water for preparation of cell culture media, Pure water for production of monoclonal antibodies, Photometry
  • 5000 MWCO microporous membrane retains macromolecules and the water can be used for cell culture applications
  • Typical applications include cell cultivation, DNA sequence analysis, IVF and general biochemical appilcations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100μS/cm
1.7 L/min.
<5 ppb
Yes
Benchtop
<1 NTU
100/240 V
Distilled, DI, RO, EDI
Type 1
Arium pro Ultrapure Water System
Cell Culture; Buffer and Media Prep; Histology; IC; Preparation of Reagents; Photometry; ELISA; AAS
<1000ppb
100/240 V