missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Benchtop Coolers
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Brand: Thermo Scientific™ 355501
232.87 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
- Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
- Holds 12, 20 or 32 tubes
- Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
- Printed grid on one-way keyed lid for proper orientation and sample identification
- Molded of impact-resistant polycarbonate with locking handle for long-term use
- Stackable to save space
Specifications
133 mm | |
226 mm | |
165 mm | |
8.94 x 6.5 x 8.25 in. (226 x 165 x 133 mm) | |
4 x 5 | |
Non-filled/clear lid | |
Benchtop Cooler | |
0.2 to 0.5 mL | |
-20°C |
8.25 in. | |
8.94 in. | |
6.5 in. | |
4x5 compartments, -20 deg. | |
<-15°C, 1 | |
Polycarbonate | |
Clear | |
20 Tubes (0.2 to 0.5 mL) |