missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Benchtop Dewar Flasks
GREENER_CHOICE

Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with these benchtop Dewar flasks with double-walled sides, insulation and pour spouts.

309.00 € - 556.00 €

Specifications

No. per Pack 1
View More Specs

Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10149270
GREENER_CHOICE
View Documents
Thermo Scientific™
4150-1000
1 L
309.00 €
Each
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
 
10693121
GREENER_CHOICE
View Documents
Thermo Scientific™
4150-2000
2 L
332.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
10782702
GREENER_CHOICE
View Documents
Thermo Scientific™
4150-4000
4 L
361.00 €
Each
In Stock
to see stock.
Add to basket
 
10007910
GREENER_CHOICE
View Documents
Thermo Scientific™
4150-9000
10 L
556.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Dewar Flasks. Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer. Flasks offer pour spouts and bottom grip area for easy removal of used liquids.

  • High density polyethylene base and HDPE cover with polyethylene coated handle for strong, shatterproof performance
  • Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer
  • Can be used from -196°C to +100°C
  • Pour spout and bottom grip area for ease of removal of used liquids
  • Vented, insulated cover allows for vapor escape but maintains sample temperature
Specifications

Specifications

1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Benchtop Dewar Flasks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.