Learn More
Wilmad-LabGlass™ Benchtop Spectrometer NMR Tubes
Wilmad-LabGlass Benchtop NMR Tubes have been tested to assure performance and permit confidence in purchasing consumables for instruments.
Brand:  Wilmad-LabGlass™ 66609-0825
 View more versions of this product
Product Code. 15806905
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 5mm O.D. tubes available in 7” or 8” lengths in quantities of 25 or 150 pieces
  • Packaging allows for easy tube access and storage
  • Attractively priced for a high-throughput laboratory environment
  • Type 1, Class B Borosilicate glass construction with disposable caps
Specifications
Specifications
Benchtop NMR Spectrometers
Borosilicate Glass
Clear
5 mm
8 in.
25
20.3 cm
NMR Tube
Cap
NMR Experiments
0.43 mm
WG-BTNMR-8-25