missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Waste Disposable Bags and Benchtop Stand

Product Code. 10631246
Click to view available options
Capacity (Metric):
2.5 L/Bag
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10631246

Brand: Ratiolab™ 7001401

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Use these bags for the disposal of contaminated labware. Waste Disposable Bags and Benchtop Stand set consists of bench-top stand and distinctive red autoclavable bags for easy identification.

  • 100 red autoclavable Bags with BIOHAZARD warning Instruction and sterilization indicator patch
  • Bags measure 210 x 290mm, made of robust polyethylene (PE)
  • Rectangle white indicator patch field turns into dark when autoclaved
  • With BIOHAZARD symbol and safety imprint in English, German, French and Spanish languages
  • Autoclavable at 121 °C, for autoclaving, the bag must not be completely closed
  • Practical bench-top stand, plastic-coated, with 3 rubber feet

Specifications

Capacity (Metric) 2.5 L/Bag
Includes 100 waste bags
Dimensions (L x W) 200 x 300mm
Product Type Table Stand and Waste Bags Set
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.