missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyflumetofen, Honeywell Fluka™
Click to view available options
Quantity:
50 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
CAS | 400882-07-7 |
Packaging | Glass Bottle |
Quantity | 50 mg |
Grade | Analytical Standard |
Boiling Point | 269°C |
For Use With (Application) | Acaricides and Insecticides |
Physical Form | Crystalline |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction