Learn More
Lenz Laborglasinstrumente™ Bent Receiver Adapter with GL Thread
Brand: Lenz Laborglasinstrumente™ 05275014
This item is not returnable.
View return policy
Specifications
Bent Receiver Adapter with GL Thread | |
Duran Glass | |
DIN 12594 | |
NS 29/32 |