missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzaldehyde, Certified AR for Analysis, Fisher Chemical
Click to view available options
250mL, Plastic Coated Glass bottle
Specifications
Specifications
CAS | 100-52-7 |
Melting Point | -26°C |
pH | 5.9 |
Boiling Point | 179°C |
Packaging | Plastic Coated Glass bottle |
Molecular Formula | C7H6O |
MDL Number | 3299 |
Quantity | 250 mL |
Formula Weight | 106.12g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Benzaldehyde
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction