missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Benzamidine HCl
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes.
93.20 € - 241.00 €
Specifications
Chemical Name or Material | Benzamidine HCl |
---|---|
CAS | 1670-14-0 |
Molecular Formula | C7H8N2-HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018066
|
Melford
B12000-100.0 |
100 g |
241.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018067
|
Melford
B12000-25.0 |
25 g |
93.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes.
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. Target enzymes include trypsin, hymotrypsin, kallikzein and plasmin.Specifications
Benzamidine HCl | |
1670-14-0 | |
C7H8N2-HCl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title