missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine Hydrochloride Hydrate, 98%

Product Code. 10549630 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10549630

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206752-36-5, 1670-14-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
SMILES Cl.NC(=N)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 86°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (on dry substance) (Argentometry)
Packaging Plastic bottle
Linear Formula HCl·xH2O
Quantity 100 g
Solubility Information Solubility in water: soluble
Formula Weight 156.61
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzamidine hydrochloride hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Benzamidine hydrochloride hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.