Learn More
Benzeneboronic acid, 98+%, Thermo Scientific Chemicals
Description
Benzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Avoid moisture exposure.
Specifications
Specifications
Melting Point | 214°C to 219°C |
Quantity | 10 g |
Beilstein | 970972 |
Sensitivity | Hygroscopic |
Merck Index | 14,1068 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 121.93 |
Percent Purity | ≥98% |
Chemical Name or Material | Benzeneboronic acid |
Safety and Handling
- Benzeneboronic acid
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- MIXTURE LIST-Contains : Benzeneboronic acid
RUO – Research Use Only