Learn More
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
38.59 € - 155.00 €
Chemical Identifiers
CAS | 98-09-9 |
---|---|
Molecular Formula | C6H5ClO2S |
Molecular Weight (g/mol) | 176.62 |
MDL Number | MFCD00007426 |
InChI Key | CSKNSYBAZOQPLR-UHFFFAOYSA-N |
Synonym | phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride |
PubChem CID | 7369 |
IUPAC Name | benzenesulfonyl chloride |
SMILES | C1=CC=C(C=C1)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10294760
|
Thermo Scientific Acros
148482500 |
250 mL | Glass bottle |
38.59 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10776191
|
Thermo Scientific Acros
148480010 |
1 L | Glass bottle |
112.56 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12153741
|
Thermo Scientific Acros
148480050 |
5 L | Plastic drum | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98-09-9 | |
176.62 | |
CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
7369 | |
C1=CC=C(C=C1)S(=O)(=O)Cl |
C6H5ClO2S | |
MFCD00007426 | |
phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
benzenesulfonyl chloride |
Specifications
98-09-9 | |
100.0 | |
1.3840g/mL | |
128°C | |
98.5% min. (GC) | |
C6H5SO2Cl | |
MFCD00007426 | |
01,46; 05,22; 14,27 | |
1.384 | |
Solubility in water: may decompose. Other solubilities: soluble in alcohol and ether | |
C1=CC=C(C=C1)S(=O)(=O)Cl | |
176.62 | |
176.62 | |
Liquid |
98.5 | |
14.5°C | |
251.0°C to 252.0°C | |
Authentic | |
C6H5ClO2S | |
1.5508 to 1.5528 | |
11,34 | |
15,1073 | |
phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
benzenesulfonyl chloride | |
7369 | |
99% | |
Benzenesulfonyl chloride, 99% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-636-6
missing translation for 'rtecsNumber' : DB8750000
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
- P363-Wash contaminated clothing before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.