missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-boronic acid, 97%, Thermo Scientific Chemicals

42.70 € - 418.00 €

Chemical Identifiers

CAS 98437-24-2
Molecular Formula C8H7BO3
Molecular Weight (g/mol) 161.95
MDL Number MFCD00236019
InChI Key PKRRNTJIHGOMRC-UHFFFAOYSA-N
Synonym benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
PubChem CID 2776266
IUPAC Name 1-benzofuran-2-ylboronic acid
SMILES OB(O)C1=CC2=CC=CC=C2O1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10413194
View Documents
Thermo Scientific Acros
378730010
1 g Glass bottle
42.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
10547624
View Documents
Thermo Scientific Acros
378730050
5 g Glass bottle
123.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
10316584
View Documents
Thermo Scientific Acros
378730250
25 g Glass bottle
418.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98437-24-2
161.95
PKRRNTJIHGOMRC-UHFFFAOYSA-N
2776266
OB(O)C1=CC2=CC=CC=C2O1
C8H7BO3
MFCD00236019
benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
1-benzofuran-2-ylboronic acid

Specifications

98437-24-2
100.0
Yellow
96% min. (HPLC)
MFCD00236019
Solubility in water: ca 2.5%
OB(O)C1=CC2=CC=CC=C2O1
161.95
161.95
Powder
96.0
112.0°C to 114.0°C
Authentic
C8H7BO3
benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
PKRRNTJIHGOMRC-UHFFFAOYSA-N
1-benzofuran-2-ylboronic acid
2776266
97%
Benzo[b]furan-2-boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

Product Identifier
  • Benzo¢b!furan-2-boronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only