missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-boronic acid, 98%, Thermo Scientific Chemicals

46.80 € - 501.00 €

Chemical Identifiers

CAS 98437-24-2
Molecular Formula C8H7BO3
Molecular Weight (g/mol) 161.95
MDL Number MFCD00236019
InChI Key PKRRNTJIHGOMRC-UHFFFAOYSA-N
Synonym benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
PubChem CID 2776266
IUPAC Name 1-benzofuran-2-ylboronic acid
SMILES OB(O)C1=CC2=CC=CC=C2O1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440088
View Documents
Thermo Scientific Alfa Aesar
B23676.03
1 g
46.80 €
1g
Please to purchase this item. Need a web account? Register with us today!
11450088
View Documents
Thermo Scientific Alfa Aesar
B23676.06
5 g
123.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11460088
View Documents
Thermo Scientific Alfa Aesar
B23676.14
25 g
501.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

Solubility
Soluble in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
Specifications

Chemical Identifiers

98437-24-2
161.95
PKRRNTJIHGOMRC-UHFFFAOYSA-N
2776266
OB(O)C1=CC2=CC=CC=C2O1
C8H7BO3
MFCD00236019
benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
1-benzofuran-2-ylboronic acid

Specifications

98437-24-2
MFCD00236019
benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
PKRRNTJIHGOMRC-UHFFFAOYSA-N
1-benzofuran-2-ylboronic acid
2776266
98%
C8H7BO3
1637889
Soluble in water.
OB(O)C1=CC2=CC=CC=C2O1
161.95
161.95
Benzo[b]furan-2-boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.