missing translation for 'onlineSavingsMsg'
Learn More
Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 10032051

Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10032051
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10032051

Brand: Thermo Scientific Acros

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-41-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00005848
InChI Key OFFSPAZVIVZPHU-UHFFFAOYSA-N
Synonym benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacidShow MoreShow Less
PubChem CID 10331
IUPAC Name 1-benzofuran-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2O1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195.0°C to 196.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 2549
Solubility Information Solubility in water: partially soluble in water,soluble in boiling wat. Other solubilities: good soluble in acetone and ethanol,slightly soluble in chloroform,carbon disulfide
Formula Weight 162.14
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Benzo[b]furan-2-carboxylic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462381 Certificate TypeCertificate of Analysis Date01/05/2024 Catalog Number233260050,233260250,233260010
Lot NumberA0432015 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number233260050,233260250,233260010
Lot NumberA0413956 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number233260050,233260250,233260010
Lot NumberA0448445 Certificate TypeCertificate of Analysis Date09/12/2022 Catalog Number233260050,233260250,233260010
Lot NumberA0421270 Certificate TypeCertificate of Analysis Date04/08/2020 Catalog Number233260050,233260250,233260010
compliance-icons
Product Identifier
  • Benzo¢b!furan-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.