missing translation for 'onlineSavingsMsg'
Learn More

Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals

Product Code. 10498430
100 mg, Glass bottle
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191-24-2
Molecular Formula C22H12
Molecular Weight (g/mol) 276.33
MDL Number MFCD00004135
InChI Key GYFAGKUZYNFMBN-UHFFFAOYSA-N
Synonym benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784
PubChem CID 9117
ChEBI CHEBI:34568
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 276.0°C to 280.0°C
Color Yellow
Boiling Point >500.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble
Formula Weight 276.33
Percent Purity 98+%
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material Benzo[ghi]perylene, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Benzo¢ghi!perylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.