missing translation for 'onlineSavingsMsg'
Learn More
Benzonitrile, for HPLC, Thermo Scientific Chemicals Product Code.: 11985641

Benzonitrile, for HPLC, Thermo Scientific Chemicals

Product Code. 11985641
1 L, Glass bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 11985641

Brand: Thermo Scientific Acros 446910010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-47-0
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.12
InChI Key JFDZBHWFFUWGJE-UHFFFAOYSA-N
Synonym phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitrilShow MoreShow Less
PubChem CID 7505
ChEBI CHEBI:27991
IUPAC Name benzonitrile
SMILES C1=CC=C(C=C1)C#N

Specifications

Melting Point -13.0°C
Density 1.0100g/mL
Boiling Point 188.0°C to 191.0°C
Flash Point 71°C
Packaging Glass bottle
Linear Formula C6H5CN
Refractive Index 1.5270 to 1.5290
Quantity 1 L
Merck Index 15,1099
Specific Gravity 1.01
Solubility Information Solubility in water: 10g/L (100°C). Other solubilities: misicble with most common organic solvents
Viscosity 1.24 mPa.s (25°C)
Formula Weight 103.12
Grade HPLC
Chemical Name or Material Benzonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461989 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number446911000,446910010
Lot NumberA0464922 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number446910010,446911000
Lot NumberA0461989 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number446910010,446911000
Lot NumberA0459285 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number446910010,446911000
Lot NumberA0452268 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog Number446910010,446911000
compliance-icons
Product Identifier
  • Benzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : Benzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzonitrile, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.