Learn More
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
23.92 € - 293.46 €
Chemical Identifiers
CAS | 119-61-9 |
---|---|
Molecular Formula | C13H10O |
Molecular Weight (g/mol) | 182.22 |
InChI Key | RWCCWEUUXYIKHB-UHFFFAOYSA-N |
Synonym | benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone |
PubChem CID | 3102 |
ChEBI | CHEBI:41308 |
IUPAC Name | diphenylmethanone |
SMILES | C1=CC=C(C=C1)C(=O)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10328740
|
Thermo Scientific Acros
219680500 |
50 g | Glass bottle |
23.92 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964851
|
Thermo Scientific Acros
219682500 |
250 g | Glass bottle |
27.84 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15129253
|
Thermo Scientific Acros
219680010 |
1 kg | Plastic bottle |
80.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15139253
|
Thermo Scientific Acros
219680050 |
5 kg | Plastic drum |
293.46 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
119-61-9 | |
182.22 | |
benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone | |
CHEBI:41308 | |
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2 |
C13H10O | |
RWCCWEUUXYIKHB-UHFFFAOYSA-N | |
3102 | |
diphenylmethanone |
Spezifikation
119-61-9 | |
White | |
143°C | |
99% min. (GC) | |
(C6H5)2CO | |
13,108 | |
benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone | |
RWCCWEUUXYIKHB-UHFFFAOYSA-N | |
diphenylmethanone | |
3102 | |
182.22 | |
Pure | |
Benzophenone |
47.0°C to 49.0°C | |
305.0°C | |
Authentic | |
C13H10O | |
07,411 | |
15,11 | |
Solubility in water: insoluble. Other solubilities: 1g/7.5mL alcohol,1g/6mL ether,soluble in chloroform,acetone and benzene | |
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2 | |
182.22 | |
CHEBI:41308 | |
99+% | |
Crystalline Powder or Flakes |
Sicherheit und Handhabung
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-337-6
- Benzophenone
Signalwort
- Danger
Gefahrenkategorie
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Gefahrenhinweis
- H350-May cause cancer.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Sicherheitshinweis
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only