missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone, 99%, pure, Thermo Scientific Chemicals

Product Code. 10529980
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
InChI Key RWCCWEUUXYIKHB-UHFFFAOYSA-N
Synonym benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
PubChem CID 3102
ChEBI CHEBI:41308
IUPAC Name diphenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Spécification

CAS Min % 98.5
CAS Max % 100.0
Melting Point 47.0°C to 49.0°C
Color White
Boiling Point 305.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5)2CO
Quantity 500 g
Beilstein 07,411
Fieser 13,108
Merck Index 15,11
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/7.5mL alcohol,1g/6mL ether,soluble in chloroform,acetone and benzene
Formula Weight 182.22
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Benzophenone
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Benzophenone
Mention d'avertissement
  • Danger
Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Mention de danger
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Conseils de prudence
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'