missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl bromide, 97%, Thermo Scientific Chemicals

25.53 € - 288.44 €

Chemical Identifiers

CAS 618-32-6
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00000112
InChI Key AQIHMSVIAGNIDM-UHFFFAOYSA-N
Synonym benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide
PubChem CID 12056
IUPAC Name benzoyl bromide
SMILES BrC(=O)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10707773
View Documents
Thermo Scientific Acros
160950050
5 mL Glass bottle
25.53 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10527563
View Documents
Thermo Scientific Acros
160950250
25 mL Glass bottle
83.82 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10570204
View Documents
Thermo Scientific Acros
160951000
100 mL Glass bottle
288.44 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

618-32-6
185.02
AQIHMSVIAGNIDM-UHFFFAOYSA-N
12056
BrC(=O)C1=CC=CC=C1
C7H5BrO
MFCD00000112
benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide
benzoyl bromide

Specifications

618-32-6
100.0
Orange
218.0°C to 219.0°C
Authentic
C7H5BrO
1.5880 to 1.5900
09,195
benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide
AQIHMSVIAGNIDM-UHFFFAOYSA-N
benzoyl bromide
12056
97%
Benzoyl bromide
96.0
-24.0°C
1.5700g/mL
90°C
96% min. (GC)
C6H5COBr
MFCD00000112
1.57
Solubility in water: may decompose. Other solubilities: miscible in ethyl ether
BrC(=O)C1=CC=CC=C1
185.02
185.02
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-544-2

Product Identifier
  • Benzoyl bromide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only