missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl chloride, 99%, pure, ACROS Organics™

Product Code. 10294650
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle

Benzoyl Chloride, 99%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00000653
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PubChem CID 7412
ChEBI CHEBI:82275
IUPAC Name benzoyl chloride
SMILES C1=CC=C(C=C1)C(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -1.0°C
Density 1.2100g/mL
Boiling Point 198.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5COCl
Refractive Index 1.5520 to 1.5540
Quantity 2.5 L
UN Number 1736
Beilstein 09, 182
Fieser 01,50; 05,24; 13,219
Merck Index 15, 1115
Specific Gravity 1.21
Solubility Information Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
Viscosity 0.0012 Pas (30°C)
Formula Weight 140.57
Percent Purity 99.5%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzoyl Chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Benzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'