missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl Chloride, Certified AR for Analysis, Fisher Chemical
Click to view available options
500mL, Plastic Coated Glass bottle
Specifications
Specifications
CAS | 98-88-4 |
Melting Point | -1°C |
Color | Colorless |
pH | 2 |
Boiling Point | 198°C |
Packaging | Plastic Coated Glass bottle |
Molecular Formula | C7H5ClO |
MDL Number | 653 |
Quantity | 500 mL |
Vapor Density | 4.88 |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction