missing translation for 'onlineSavingsMsg'
Learn More
Benzyl 2,3-O-isopropylidene-alpha-D-mannofuranoside, Thermo Scientific™ Product Code.: 15457539

Benzyl 2,3-O-isopropylidene-alpha-D-mannofuranoside, Thermo Scientific™

Product Code. 15457539
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15457539

Brand: Thermo Scientific Alfa Aesar J66254.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in chloroform, dichloromethane or DMSO

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 20689-03-6
Color White
Molecular Formula C16H22O6
MDL Number MFCD02683277
Quantity 1 g
Synonym benzyl 2,3-o-isopropylidene-alpha-d-mannofuranoside, benzyl 2,3-o-isopropylidene-, a-d-mannofuranoside, phenylmethyl 2,3-o-1-methylethylidene-, benzyl 3xi-2,3-o-1-methylethylidene-alpha-d-erythro-hexofuranoside, 1r-1-4r,6s,6ar-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl ethane-1,2-diol
Solubility Information Soluble in chloroform, dichloromethane or DMSO
InChI Key NKCNUNUEQRYLNZ-MLQJFRMTSA-N
SMILES CC1(OC2C(O1)C(OC2C(CO)O)OCC3=CC=CC=C3)C
IUPAC Name (1R)-1-[(3aR,4S,6R)-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]ethane-1,2-diol
Molecular Weight (g/mol) 310.346
PubChem CID 57370143
Formula Weight 310.34
Physical Form Powder
Chemical Name or Material Benzyl 2,3-O-isopropylidene-alpha-D-mannofuranoside
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ17B024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberZ16A041 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl 2,3-O-isopropylidene-alpha-D-mannofuranoside, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.