missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 3-bromopropyl ether, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Density | 1.2980g/mL |
| Boiling Point | 130°C to 132°C (8.0 mmHg) |
| Flash Point | 113°C |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 10 g |
| Specific Gravity | 1.298 |
| Formula Weight | 229.11 |
| Show More |
Safety and Handling
Product Identifier
- Benzyl 3-bromopropyl ether
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction