Learn More
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals
32.16 € - 98.79 €
Chemical Identifiers
CAS | 5396-89-4 |
---|---|
Molecular Formula | C11H12O3 |
Molecular Weight (g/mol) | 192.214 |
MDL Number | MFCD00008785 |
InChI Key | WOFAGNLBCJWEOE-UHFFFAOYSA-N |
Synonym | benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x |
PubChem CID | 142266 |
IUPAC Name | benzyl 3-oxobutanoate |
SMILES | CC(=O)CC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304589
|
Thermo Scientific Alfa Aesar
A14915.14 |
25 g |
32.16 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452727
|
Thermo Scientific Alfa Aesar
A14915.18 |
50 g |
45.02 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314589
|
Thermo Scientific Alfa Aesar
A14915.22 |
100 g |
78.59 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11462727
|
Thermo Scientific Alfa Aesar
A14915.30 |
250 g |
98.79 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.
Solubility
It is soluble in alkali solutions.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
5396-89-4 | |
192.214 | |
WOFAGNLBCJWEOE-UHFFFAOYSA-N | |
142266 | |
CC(=O)CC(=O)OCC1=CC=CC=C1 |
C11H12O3 | |
MFCD00008785 | |
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x | |
benzyl 3-oxobutanoate |
Specifications
5396-89-4 | |
276°C | |
C11H12O3 | |
MFCD00008785 | |
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x | |
WOFAGNLBCJWEOE-UHFFFAOYSA-N | |
benzyl 3-oxobutanoate | |
142266 | |
97% |
1.113 | |
108°C (226°F) | |
1.512 | |
2049615 | |
It is soluble in alkali solutions. | |
CC(=O)CC(=O)OCC1=CC=CC=C1 | |
192.214 | |
192.22 | |
Benzyl acetoacetate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'einecsNumber' : 226-416-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.