missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Preparation of high refractive index polyacrylates.Benzyl acrylate is used in the preparation of heptanoic acid benzyl ester. It is used to prepare polybenzylacrylate using azobisisobutyronitrile as initiator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Density | 1.06 g/mL |
Boiling Point | 111°C (8 mmHg) |
Linear Formula | C6H5CH2CO2CH=CH2 |
Refractive Index | 1.518 |
Quantity | 250 g |
UN Number | UN3082 |
Solubility Information | Miscible with organic solvents and water. |
Formula Weight | 162.19 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'