missing translation for 'onlineSavingsMsg'
Learn More
Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol, Thermo Scientific Chemicals Product Code.: 11369268

Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11369268
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11369268

Brand: Thermo Scientific Alfa Aesar 043204.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preparation of high refractive index polyacrylates

Preparation of high refractive index polyacrylates.Benzyl acrylate is used in the preparation of heptanoic acid benzyl ester. It is used to prepare polybenzylacrylate using azobisisobutyronitrile as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2495-35-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00048147
InChI Key GCTPMLUUWLLESL-UHFFFAOYSA-N
Synonym benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl
PubChem CID 75617
IUPAC Name benzyl prop-2-enoate
SMILES C=CC(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Density 1.06 g/mL
Boiling Point 111°C (8 mmHg)
Linear Formula C6H5CH2CO2CH=CH2
Refractive Index 1.518
Quantity 50 g
UN Number UN3082
Solubility Information Miscible with organic solvents and water.
Formula Weight 162.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzyl acrylate, Stabilized with ≈150ppm 4-methoxyphenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS25F058 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043204.18,043204.30,043204.09
Lot NumberA22T031 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043204.18,043204.30,043204.09
Lot NumberF22X044 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043204.18,043204.30,043204.09
Lot NumberB18P09 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043204.18,043204.30,043204.09
Lot NumberE06U016 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043204.18,043204.30,043204.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.