missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl alcohol, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 100-51-6 |
Melting Point | -15.0°C |
Density | 1.0440g/mL |
Boiling Point | 205.0°C |
Flash Point | 96°C |
Packaging | Glass Bottle |
Molecular Formula | C7H8O |
Linear Formula | C6H5CH2OH |
Refractive Index | 1.5380 to 1.5410 |
MDL Number | MFCD00004599,MFCD03792087 |
Show More |
Safety and Handling
Product Identifier
- Benzyl alcohol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'