missing translation for 'onlineSavingsMsg'
Learn More
Benzyl alcohol, specified according to requirements of Ph.Eur., Thermo Scientific Chemicals Product Code.: 12695037

Benzyl alcohol, specified according to requirements of Ph.Eur., Thermo Scientific Chemicals

Product Code. 12695037
1 L, Glass bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 12695037

Brand: Thermo Scientific Acros 447000010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -15.0°C
Density 1.0440g/mL
Boiling Point 205.0°C
Flash Point 96°C
Packaging Glass bottle
Linear Formula C6H5CH2OH
Refractive Index 1.5380 to 1.5410
Quantity 1 L
Merck Index 15,1127
Specific Gravity 1.044
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride
Viscosity 6.6 mPa.s (20°C)
Formula Weight 108.14
Related Substances greater than that of benzylalcohol)
Grade ACS Reagent
Chemical Name or Material Benzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl alcohol, specified according to requirements of Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.