missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl benzoate, 99+%
Alle ansehen Thermo Scientific Chemicals Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
missing translation for 'unitSize'
1 L
250mL
5 L
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Acidity | 0.3mg KOH/g max. |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | 18.0°C to 20.0°C |
| Color | Colorless |
| Density | 1.1180g/mL |
| Boiling Point | 323.0°C |
| Flash Point | 147°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (GC) |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Benzyl benzoate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur