missing translation for 'onlineSavingsMsg'
Learn More

Benzyl bromide, 98%

Product Code. 10767583 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 10767583

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-39-0 | C7H7Br | 171.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-39-0
MDL Number MFCD00000172
InChI Key AGEZXYOZHKGVCM-UHFFFAOYSA-N
Synonym benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
PubChem CID 7498
ChEBI CHEBI:59858
IUPAC Name bromomethylbenzene
SMILES C1=CC=C(C=C1)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2°C
Color Colorless to Yellow
Density 1.4300g/mL
Boiling Point 198.0°C to 199.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7Br
Linear Formula C6H5CH2Br
Refractive Index 1.5742 to 1.5762
Quantity 1 L
Beilstein 05, 306
Fieser 05,25; 13,51
Merck Index 15, 1131
Specific Gravity 1.43
Solubility Information Solubility in water: 436 mg/l (25°C). Other solubilities: slowly decomposes, soluble in alcohol, benzene and ether
Molecular Weight (g/mol) 171.04
Formula Weight 171.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzyl bromide, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.