missing translation for 'onlineSavingsMsg'
Learn More
Benzyl carbamate, 99%, Thermo Scientific Chemicals Product Code.: 11474623

Benzyl carbamate, 99%, Thermo Scientific Chemicals

Product Code. 11474623
25 g
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy

Product Code. 11474623

Brand: Thermo Scientific Alfa Aesar A11569.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-84-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007965
InChI Key PUJDIJCNWFYVJX-UHFFFAOYSA-N
Synonym carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
PubChem CID 12136
IUPAC Name benzyl carbamate
SMILES NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 85°C to 89°C
Beilstein 1865635
Quantity 25 g
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material Benzyl carbamate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA24N31 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11569.14,A11569.22,A11569.30
Lot Number5015F04W Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11569.14,A11569.22,A11569.30
Lot NumberG1970A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11569.14,A11569.22,A11569.30
Lot Number10212500 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11569.14,A11569.22,A11569.30
Lot Number10233892 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11569.14,A11569.22,A11569.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl carbamate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.