Learn More
Benzyl chloride, 99%, stab., Thermo Scientific Chemicals
25.73 € - 244.22 €
Chemical Identifiers
CAS | 100-44-7 |
---|---|
Molecular Formula | C7H7Cl |
Molecular Weight (g/mol) | 126.583 |
MDL Number | MFCD00000889 |
InChI Key | KCXMKQUNVWSEMD-UHFFFAOYSA-N |
Synonym | benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluolShow More |
PubChem CID | 7503 |
ChEBI | CHEBI:615597 |
IUPAC Name | chloromethylbenzene |
SMILES | C1=CC=C(C=C1)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11464094
|
Thermo Scientific Alfa Aesar
A12481.36 |
500 g |
25.73 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11474094
|
Thermo Scientific Alfa Aesar
A12481.0E |
2500 g |
78.89 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11484094
|
Thermo Scientific Alfa Aesar
A12481.0C |
10,000 g | N/A | N/A | N/A | |||||
Description
Benzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.
Solubility
Miscible with alcohol, ether. Insoluble in waterMiscible with alcohol, ether and chloroform.
Notes
Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with common metals, lead,oxidizing agents, iron, iron salts, brass and aluminum
Chemical Identifiers
100-44-7 | |
126.583 | |
KCXMKQUNVWSEMD-UHFFFAOYSA-N | |
7503 | |
chloromethylbenzene |
C7H7Cl | |
MFCD00000889 | |
benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluolShow More | |
CHEBI:615597 | |
C1=CC=C(C=C1)CCl |
Specifications
100-44-7 | |
1.099 | |
73°C (163°F) | |
C7H7Cl | |
MFCD00000889 | |
471308 | |
benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol | |
KCXMKQUNVWSEMD-UHFFFAOYSA-N | |
chloromethylbenzene | |
7503 | |
126.59 | |
Liquid |
°C to 43°C | |
178°C to 180°C | |
99%, Stabilized | |
1.538 | |
UN1738 | |
14,1129 | |
Miscible with alcohol,ether; Insoluble in waterMiscible with alcohol,ether and chloroform. | |
C1=CC=C(C=C1)CCl | |
126.583 | |
CHEBI:615597 | |
99% | |
Benzyl Chloride, stabilized |
Safety and Handling
GHS H Statement
H301-H330-H350-H373-H314-H318-H227-H335
P201-P202-P210-P234-P235-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P333+P313-P362-P370+P378q-P390-P501c
H227-H290-H302-H315-H317-H318-H331-H335-H336-H340-H350-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZYL CHLORIDE
missing translation for 'einecsNumber' : 202-853-6
missing translation for 'rtecsNumber' : XS8925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Benzyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Benzyl chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.