missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroformate, 97 wt%, stabilized, Thermo Scientific Chemicals

Product Code. 10183580
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Benzyl chloroformate, 97 wt%, stabilized, Acros Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 178.0 deg.C to 180.0 deg.C, Undesignated, Melting Point: -20.0 deg.C, Molecular Weight: 170.6, Percent Purity: 97 wt%, Assay Percent Range: 96% min. (GC), Beilstein: 06,437

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-53-1, 100-44-7
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
PubChem CID 10387
IUPAC Name benzyl carbonochloridate
SMILES C1=CC=C(C=C1)COC(=O)Cl

Specifications

Melting Point -20.0°C
Density 1.2000g/mL
Boiling Point 178.0°C to 180.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2CH2C6H5
Refractive Index 1.5160 to 1.5180
Quantity 5 g
Beilstein 06,437
Fieser 01,109; 02,59; 15,22
Merck Index 15,1801
Specific Gravity 1.2
Solubility Information Solubility in water: reacts
Stabilizer 0.1% anhydrous sodium carbonate
Viscosity 2.36 mPa.s (20°C)
Formula Weight 170.6
Percent Purity 97 wt%
Physical Form Liquid
Chemical Name or Material Benzyl chloroformate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyl chloroformate, stabilized
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Benzyl chloroformate, benzyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.