missing translation for 'onlineSavingsMsg'
Learn More

Benzyl n-butyl ether, 97%, Thermo Scientific Chemicals

Product Code. 11382597
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 588-67-0
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00039954
InChI Key MAYUYFCAPVDYBQ-UHFFFAOYSA-N
Synonym benzyl butyl ether, ether, benzyl butyl, butyl benzyl ether, benzyl n-butyl ether, benzene, butoxymethyl, butoxymethyl benzene, n-butyl benzyl ether, unii-8483x8zo8n, ether, benzyl n-butyl, butoxyphenylmethane
PubChem CID 61134
IUPAC Name butoxymethylbenzene
SMILES CCCCOCC1=CC=CC=C1

Specifications

Density 0.92
Boiling Point 111°C to 112°C (23 mmHg)
Refractive Index 1.491
Quantity 5 g
Beilstein 1935234
Formula Weight 164.25
Percent Purity 97%
Chemical Name or Material Benzyl n-butyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.