missing translation for 'onlineSavingsMsg'
Learn More

Benzyl salicylate, 99%, Thermo Scientific Chemicals

Product Code. 11473257
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-58-1
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00020034
InChI Key ZCTQGTTXIYCGGC-UHFFFAOYSA-N
Synonym benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester
PubChem CID 8363
IUPAC Name benzyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O

Specifications

Melting Point 18°C to 20°C
Density 1.17
Boiling Point 190°C (14 mmHg)
Flash Point 137°C (278°F)
Refractive Index 1.5804
Quantity 2500 g
Beilstein 2115365
Merck Index 14,1144
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Benzyl salicylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.