missing translation for 'onlineSavingsMsg'
Learn More

Benzylmagnesium chloride, 1.5M solution in THF, Thermo Scientific Chemicals

80.40 € - 268.00 €

Chemical Identifiers

CAS 6921-34-2, 109-99-9
Molecular Formula C7H7ClMg
Molecular Weight (g/mol) 150.89
MDL Number MFCD00000469
InChI Key NRAFPLGJPPJUNB-UHFFFAOYSA-M
Synonym benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium
PubChem CID 2733352
IUPAC Name benzyl(chloro)magnesium
SMILES Cl[Mg]CC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10095093
View Documents Promotion Details
Thermo Scientific Acros
389551000
100 mL AcroSeal™ Glass bottle
80.40 €
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10463735
View Documents Promotion Details
Thermo Scientific Acros
389558000
800 mL AcroSeal™ Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6921-34-2, 109-99-9
150.89
NRAFPLGJPPJUNB-UHFFFAOYSA-M
2733352
Cl[Mg]CC1=CC=CC=C1
C7H7ClMg
MFCD00000469
benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium
benzyl(chloro)magnesium

Specifications

Brown to Black
20.51, 75.83
C7H7ClMg
MFCD00000469
1.0300g/mL
Solubility in water: reacts.
Cl[Mg]CC1=CC=CC=C1
150.89
150.89
1.03
Iridescent Powder or Crystals
6921-34-2, 109-99-9
24.17, 79.49
(C6H5)CH2MgCl
1.40 to 1.65M
benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium
NRAFPLGJPPJUNB-UHFFFAOYSA-M
benzyl(chloro)magnesium
2733352
19 to 21% (as BMC) (Total base)
−17°C
Benzylmagnesium chloride, 1.5M solution in THF
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only