missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltri-n-butylammonium bromide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
250 g
Description
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature.
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature.
Specifications
Specifications
Melting Point | ∼175°C (decomposition) |
Quantity | 25 g |
Beilstein | 3776294 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 356.4 |
Percent Purity | 99% |
Chemical Name or Material | Benzyltri-n-butylammonium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'