missing translation for 'onlineSavingsMsg'
Learn More

Benzyltri-n-butylammonium bromide, 99%, Thermo Scientific Chemicals

Product Code. 11416816
Click to view available options
Quantity:
25 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-59-0
Molecular Formula C19H34BrN
Molecular Weight (g/mol) 356.392
MDL Number MFCD00011848
InChI Key UDYGXWPMSJPFDG-UHFFFAOYSA-M
Synonym benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
PubChem CID 2724282
IUPAC Name benzyl(tributyl)azanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]

Specifications

Melting Point ∼175°C (decomposition)
Quantity 250 g
Beilstein 3776294
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 356.4
Percent Purity 99%
Chemical Name or Material Benzyltri-n-butylammonium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'