missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals

Product Code. 11442868
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5350-41-4
Molecular Formula C10H16BrN
Molecular Weight (g/mol) 230.149
MDL Number MFCD00011780
InChI Key UUZYBYIOAZTMGC-UHFFFAOYSA-M
Synonym benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm
PubChem CID 21449
IUPAC Name benzyl(trimethyl)azanium;bromide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[Br-]

Specifications

Melting Point 228°C to 232°C
Quantity 25 g
Beilstein 3917251
Sensitivity Hygroscopic
Formula Weight 230.15
Percent Purity ≥98%
Chemical Name or Material Benzyltrimethylammonium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'