missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium chloride, 50 wt.% solution in water, Thermo Scientific Chemicals

Product Code. 10430871
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-93-9, 7732-18-5
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00011782
InChI Key KXHPPCXNWTUNSB-UHFFFAOYSA-M
Synonym benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
PubChem CID 5963
IUPAC Name benzyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC1=CC=CC=C1

Specifications

Color Colorless to Yellow
CAS Min % 49.0
CAS Max % 51.0
Linear Formula C6H5CH2N(CH3)3Cl
Beilstein 12, 1020
Fieser 01,53
Density 1.0440g/mL
Formula Weight 185.7
Percent Purity ≥49%
Packaging Glass bottle
Refractive Index 1.445
Specific Gravity 1.044
Quantity 100 g
Physical Form Liquid
Chemical Name or Material Benzyltrimethylammonium chloride, 50 wt. % Solution in Water
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'