missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium dichloroiodate, 95%, Thermo Scientific Chemicals

Product Code. 11449947
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114971-52-7
Molecular Formula C10H16Cl2IN
Molecular Weight (g/mol) 348.049
MDL Number MFCD00075259
InChI Key PPDJNZTUDFPAHX-UHFFFAOYSA-N
Synonym benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
PubChem CID 2724680
SMILES C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl

Specifications

Melting Point ∼125°C
Linear Formula C6H5CH2(CH3)3NICl2
Quantity 25 g
Beilstein 4611949
Sensitivity Hygroscopic; Light sensitive
Formula Weight 348.06
Percent Purity 95%
Chemical Name or Material Benzyltrimethylammonium dichloroiodate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'