missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltrimethylammonium hydroxide, 40 wt% solution in methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Color | Yellow |
CAS Min % | 39.0, 59.0 |
CAS Max % | 41.0, 61.0 |
Linear Formula | C6H5CH2N(CH3)3OH |
Fieser | 01,1252; 05,29; 08,36; 12,48; 05,30 |
Density | 0.9250g/mL |
Solubility Information | Solubility in water: miscible. |
Formula Weight | 167.25 |
Percent Purity | ≥40% |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Benzyltrimethylammonium hydroxide, 40 wt% solution in methanol
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after single exposure Category 1
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'