Learn More
Beperidium iodide, MedChemExpress
MedChemExpress Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Brand:  Medchem Express HY-100152
Encompass_Preferred
 View more versions of this product
Product Code. 16465547
  • 544.05 € / 5mg
Explore more special offers
Description
Description
Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Specifications
Specifications
Beperidium iodide
White
C23H34IN3O3
SX 810
C[N+]1(CC)CCC(OC(C(C2=NOC3=C2C=CC=C3)N4CCCCCC4)=O)CC1.[I-]
527.44
Research
COVID-19-immunoregulation
86434-57-3
Research
5 mg
DMSO : ≥ 100 mg/mL (189.60 mM)
527.44
99.79%
Solid