Learn More
Berkshire™ Gamma Wipe™ 67
BIOPHARMA
Berkshire Gamma Wipe™ 67 is a polyester-cellulose blend wiper recommended for ISO Class 5 and above environments
Brand:  Berkshire™ GW67ST0980
Product Code. 15622329
  • 856.00 € / Pack of 1600
Explore more special offers
Description
Description
  • Smooth, highly absorbent and durable with low extractable levels
  • Economical
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • 45% polyester and 55% cellulose
  • Each package is gamma irradiated to a 10-6 Sterility Assurance
Specifications
Specifications
9 in.
9 in.
Non-woven Polyester Cellulose
Designed for use in ISO Class 5 and higher sterile environments and USP <797> applications
23 cm
23 cm
White
Sterile