missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ MicroSeal™ 1200 Wipers
BIOPHARMA

For ISO Class 3 (Class 1) applications

Brand:  Berkshire™ MS1200.1212.6

 View more versions of this product

Product Code. 17414302

  • 1125.00 € / Pack of 600
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

All-purpose wipers for use in spill control, equipment cleaning, and general cleanup.

  • Made of ultrasonically sealed edge 100% continuous filament polyester knit
  • Works well with IPA, acetone, and other solvents
  • Laundered and packed in an ISO 4 (Class 10) cleanroom
  • Critically low particle and fiber generation

† Bulk packed.

Specifications

Specifications

Cleanroom Wipe
30.48 cm
30.48 cm
IPA, Acetone and other solvents
12 x 12 in. (30.48 x 30.48 cm)
Photolithography Process areas, flat panel, optics, and chamber cleans
12 in.
12 in.
Polyester Knit
Sealed
For use in ISO Class 3, spill control, equipment cleaning and general cleanup
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.