missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ ValuSeal-LP™ Wipers - ISO Class 3
BIOPHARMA

For ISO 3 (Class 1) cleanroom applications

Brand:  Berkshire™ VSLP.1212.14

 View more versions of this product

Product Code. 15533114

  • 1935.00 € / Pack of 14
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from 100% continuous filament polyester knit
  • Ultrasonically sealed edge for reduced fiber contamination
  • Abrasion resistant with critically low particles and fibers and low non-volatile residues
  • Chemically compatible with IPA, acetone and other solvents
  • Steam autoclavable
  • Laundered and packed in an ISO 4 (Class 10) cleanroom

†Bulk packed.

Order Info

Basis weight, 145g/m2

Specifications

Specifications

Cleanroom Wipe
30.48 cm
30.48 cm
IPA, Acetone and other solvents
Sealed
For ISO 3 highest level of contamination control in critical processing applications
1050 Sheets/Cs.
12 in.
12 in.
Polyester Knit
Autoclavable
12 x 12 in. (30.48 x 30.48 cm)
For cleaning of medical device products
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.