missing translation for 'onlineSavingsMsg'
Learn More

Beryllium foil, 0.05mm (0.002in) thick, 99.5% (metals basis), Thermo Scientific Chemicals

724.61 € - 1915.00 €

Chemical Identifiers

CAS 7440-41-7
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11308758
SDS View Documents
Thermo Scientific Alfa Aesar
041613.FF
25 x 25 mm
724.61 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11410411
SDS View Documents
Thermo Scientific Alfa Aesar
041613.FI
50 x 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Not vacuum tight
Specifications

Chemical Identifiers

7440-41-7
9.01
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
5460467
beryllium
Be
MFCD00134032
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
CHEBI:33783
[Be]

Specifications

7440-41-7
MFCD00134032
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
beryllium
5460467
99.5%
≈0.058g/25x25mm
(metals basis)
Be
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
[Be]
9.01
CHEBI:33783
Odorless
Foil
Beryllium foil, 0.05mm (0.002 in.) thick, Not certified pinhole free
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c

H301-H315-H317-H319-H330-H335-H350i-H372

missing translation for 'einecsNumber' : 231-150-7

missing translation for 'rtecsNumber' : DS1750000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Not vacuum tight

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.